Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108094.2 | NP_001101564.1 | 5 - 6 | 735 | 156 | |
GenBank mRNA | BC128768.1 | - | 5 - 6 | 735 | 7125536 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |