Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012619.2 | NP_036751.2 | 6 - 7 | 794 | 78 | |
XM_008765215.1 | XP_008763437.1 | 4 - 5 | 734 | 78 | ||
GenBank mRNA | BC078881.1 | - | 6 - 7 | 794 | 7131414 | 78 |
FQ219397.1 | - | 2 - 3 | 230 | 78 | ||
K02599.1 | - | 1 - 2 | 84 | 78 | ||
M12337.1 | - | 6 - 7 | 779 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |