Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017313.3 | NP_059009.2 | 1 - 2 | 282 | 141 | |
XM_006241357.2 | XP_006241419.1 | 2 - 3 | 451 | 141 | ||
XM_006241358.2 | XP_006241420.1 | 2 - 3 | 456 | 141 | ||
XM_006241359.2 | XP_006241421.1 | 2 - 3 | 1460 | 141 | ||
XM_006241360.2 | XP_006241422.1 | 2 - 3 | 446 | 141 | ||
XM_008765382.1 | XP_008763604.1 | 1 - 2 | 508 | 141 | ||
GenBank mRNA | BC127500.1 | - | 1 - 2 | 285 | 7457420 | 141 |
U19181.1 | - | 1 - 2 | 275 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |