Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053729.1 | NP_446181.1 | 5 - 6 | 657 | 126 | |
GenBank mRNA | AY026945.1 | - | 5 - 6 | 657 | 126 | |
BC129088.1 | - | 5 - 6 | 674 | 8362408 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |