Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241616.1 | XP_006241678.1 | 5 - 6 | 703 | 105 | |
XM_006241618.2 | XP_006241680.1 | 4 - 5 | 863 | 105 | ||
XM_006241619.2 | XP_006241681.1 | 2 - 3 | 1630 | 105 | ||
XM_008765476.1 | XP_008763698.1 | 4 - 5 | 644 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |