Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241432.2 | XP_006241494.1 | 2 - 3 | 580 | 76 | |
XM_006241433.2 | XP_006241495.1 | 3 - 4 | 2096 | 76 | ||
XM_006241434.2 | XP_006241496.1 | 3 - 4 | 447 | 76 | ||
XM_006241435.2 | XP_006241497.1 | 2 - 3 | 2060 | 76 | ||
GenBank mRNA | X97445.1 | - | 2 - 3 | 163 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |