Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130561.2 | NP_001124033.1 | 4 - 5 | 694 | 145 | |
XM_006241614.2 | XP_006241676.1 | 4 - 5 | 793 | 145 | ||
GenBank mRNA | BC169014.1 | - | 4 - 5 | 694 | 9091867 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |