Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024398.2 | NP_077374.2 | 16 - 17 | 2090 | 121 | |
GenBank mRNA | AJ243266.1 | - | 16 - 17 | 2091 | 121 | |
BC061999.1 | - | 16 - 17 | 2090 | 5621226 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |