Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009665.1 | NP_001009665.1 | 3 - 4 | 374 | 83 | |
XM_006241613.2 | XP_006241675.1 | 3 - 4 | 749 | 83 | ||
XM_008765451.1 | XP_008763673.1 | 2 - 3 | 462 | 83 | ||
XM_008765452.1 | XP_008763674.1 | 3 - 4 | 3412 | 83 | ||
GenBank mRNA | BC087093.1 | - | 3 - 4 | 374 | 7191226 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |