Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130538.1 | NP_001124010.1 | 2 - 3 | 574 | 82 | |
XM_006241645.2 | XP_006241707.1 | 2 - 3 | 574 | 82 | ||
XM_008765500.1 | XP_008763722.1 | 2 - 3 | 331 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |