Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130576.1 | NP_001124048.1 | 2 - 3 | 811 | 97 | |
XM_008765518.1 | XP_008763740.1 | 3 - 4 | 3446 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |