Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765525.1 | XP_008763747.1 | 6 - 7 | 1112 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |