Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130545.1 | NP_001124017.1 | 1 - 2 | 131 | 60 | |
XM_006241726.2 | XP_006241788.1 | 1 - 2 | 319 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |