Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199110.1 | NP_954541.1 | 6 - 7 | 914 | 89 | |
GenBank mRNA | BC061801.1 | - | 6 - 7 | 914 | 5598335 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |