Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135913.1 | NP_001129385.1 | 2 - 3 | 546 | 109 | |
XM_006242122.2 | XP_006242184.1 | 3 - 4 | 1043 | 109 | ||
XM_008765787.1 | XP_008764009.1 | 3 - 4 | 1043 | 109 | ||
XM_008765788.1 | XP_008764010.1 | 3 - 4 | 1043 | 109 | ||
GenBank mRNA | BC167764.1 | - | 2 - 3 | 546 | 7453894 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |