Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025659.2 | NP_001020830.2 | 1 - 2 | 77 | 74 | |
GenBank mRNA | BC097394.1 | - | 1 - 2 | 77 | 7453303 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |