Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031122.1 | NP_112384.1 | 6 - 7 | 531 | 65 | |
GenBank mRNA | BC078804.1 | - | 6 - 7 | 538 | 7109473 | 65 |
FQ212434.1 | - | 588 | 65 | |||
FQ228746.1 | - | 6 - 7 | 493 | 65 | ||
X82021.1 | - | 6 - 7 | 531 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |