Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134765.1 | NP_001128237.1 | 2 - 3 | 257 | 88 | |
XM_008765519.1 | XP_008763741.1 | 15 - 16 | 1774 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |