Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114405.2 | NP_001107877.1 | 2 - 3 | 691 | 53 | |
XM_006241729.2 | XP_006241791.1 | 4 - 5 | 1188 | 53 | ||
XM_006241731.2 | XP_006241793.1 | 3 - 4 | 2298 | 53 | ||
XM_006241732.2 | XP_006241794.1 | 3 - 4 | 1110 | 53 | ||
XM_006241733.2 | XP_006241795.1 | 3 - 4 | 1069 | 53 | ||
XM_008765573.1 | XP_008763795.1 | 4 - 5 | 2376 | 53 | ||
XM_008765574.1 | XP_008763796.1 | 3 - 4 | 1755 | 53 | ||
GenBank mRNA | BC159432.1 | - | 2 - 3 | 691 | 9092581 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |