Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017142.1 | NP_058838.1 | 16 - 17 | 3917 | 123 | |
XM_006241703.2 | XP_006241765.1 | 15 - 16 | 4320 | 123 | ||
XM_006241704.2 | XP_006241766.1 | 15 - 16 | 4212 | 123 | ||
GenBank mRNA | L26986.1 | - | 16 - 17 | 3917 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |