Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013219.1 | NP_001013237.1 | 4 - 5 | 721 | 86 | |
XM_006241709.2 | XP_006241771.1 | 7 - 8 | 1693 | 86 | ||
XM_006241710.2 | XP_006241772.1 | 7 - 8 | 2611 | 86 | ||
XM_008765513.1 | XP_008763735.1 | 6 - 7 | 1638 | 86 | ||
GenBank mRNA | AJ748840.1 | - | 4 - 5 | 721 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |