Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013948.1 | NP_001013970.1 | 3 - 4 | 514 | 114 | |
XM_006241530.2 | XP_006241592.1 | 3 - 4 | 785 | 114 | ||
XM_006241531.2 | XP_006241593.1 | 2 - 3 | 417 | 114 | ||
XM_006241532.2 | XP_006241594.1 | 2 - 3 | 2158 | 114 | ||
XM_006241533.2 | XP_006241595.1 | 2 - 3 | 2084 | 114 | ||
GenBank mRNA | AB095364.1 | - | 3 - 4 | 514 | 114 | |
BC105616.1 | - | 3 - 4 | 510 | 7453107 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |