Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013104.1 | NP_001013122.1 | 6 - 7 | 1719 | 70 | |
XM_008765549.1 | XP_008763771.1 | 7 - 8 | 3840 | 70 | ||
XM_008765550.1 | XP_008763772.1 | 7 - 8 | 1901 | 70 | ||
XM_008765551.1 | XP_008763773.1 | 9 - 10 | 1893 | 70 | ||
XM_008765552.1 | XP_008763774.1 | 9 - 10 | 1874 | 70 | ||
XM_008765553.1 | XP_008763775.1 | 7 - 8 | 1793 | 70 | ||
XM_008765554.1 | XP_008763776.1 | 6 - 7 | 1847 | 70 | ||
XM_008765555.1 | XP_008763777.1 | 6 - 7 | 1832 | 70 | ||
XM_008765556.1 | XP_008763778.1 | 5 - 6 | 1775 | 70 | ||
XM_008765557.1 | XP_008763779.1 | 6 - 7 | 723 | 70 | ||
XM_008765558.1 | XP_008763780.1 | 6 - 7 | 730 | 70 | ||
XM_008765559.1 | XP_008763781.1 | 6 - 7 | 710 | 70 | ||
XM_008765560.1 | XP_008763782.1 | 6 - 7 | 708 | 70 | ||
XM_008765561.1 | XP_008763783.1 | 5 - 6 | 651 | 70 | ||
XM_008765562.1 | XP_008763784.1 | 5 - 6 | 636 | 70 | ||
GenBank mRNA | BC079391.1 | - | 6 - 7 | 1719 | 7111852 | 70 |
FQ220813.1 | - | 6 - 7 | 680 | 70 | ||
FQ225933.1 | - | 6 - 7 | 715 | 70 | ||
FQ227443.1 | - | 6 - 7 | 691 | 70 | ||
FQ233760.1 | - | 6 - 7 | 671 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |