Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191114.1 | NP_001178043.1 | 2 - 3 | 95 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |