Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009292.1 | NP_001009292.1 | 2 - 3 | 362 | 91 | |
GenBank mRNA | BC087090.1 | - | 2 - 3 | 362 | 7190632 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |