Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079894.1 | NP_001073363.1 | 9 - 10 | 870 | 140 | |
XM_006230409.2 | XP_006230471.1 | 9 - 10 | 948 | 140 | ||
XM_006230412.2 | XP_006230474.1 | 9 - 10 | 949 | 140 | ||
XM_006230413.2 | XP_006230475.1 | 9 - 10 | 948 | 140 | ||
XM_006230415.2 | XP_006230477.1 | 9 - 10 | 948 | 140 | ||
XM_008759907.1 | XP_008758129.1 | 9 - 10 | 1023 | 140 | ||
XM_008759908.1 | XP_008758130.1 | 9 - 10 | 1344 | 140 | ||
XM_008759909.1 | XP_008758131.1 | 9 - 10 | 917 | 140 | ||
XM_008759910.1 | XP_008758132.1 | 6 - 7 | 587 | 140 | ||
GenBank mRNA | BC128752.1 | - | 9 - 10 | 870 | 7100072 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |