Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107552.1 | NP_001101022.1 | 5 - 6 | 1072 | 134 | |
XM_006230276.2 | XP_006230338.1 | 7 - 8 | 1768 | 134 | ||
XM_006230277.2 | XP_006230339.1 | 7 - 8 | 1639 | 134 | ||
XM_006230278.2 | XP_006230340.1 | 6 - 7 | 1549 | 134 | ||
XM_006230279.2 | XP_006230341.1 | 6 - 7 | 1420 | 134 | ||
XM_006230280.2 | XP_006230342.1 | 7 - 8 | 1652 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |