Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017046.1 | NP_058742.1 | 8 - 9 | 1392 | 84 | |
GenBank mRNA | EU627517.1 | - | 7 - 8 | 1294 | 84 | |
U37539.1 | - | 8 - 9 | 1346 | 84 | ||
U37540.1 | - | 8 - 9 | 1346 | 84 | ||
X77933.1 | - | 8 - 9 | 1392 | 84 | ||
X78034.1 | - | 8 - 9 | 1375 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |