Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030853.1 | NP_110480.1 | 4 - 5 | 487 | 119 | |
XM_008759919.1 | XP_008758141.1 | 4 - 5 | 941 | 119 | ||
GenBank mRNA | AJ001184.1 | - | 4 - 5 | 487 | 119 | |
BC088130.1 | - | 4 - 5 | 447 | 7382238 | 119 | |
FQ233731.1 | - | 4 - 5 | 504 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |