Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034911.1 | NP_001030083.1 | 4 - 5 | 641 | 96 | |
XM_006230189.2 | XP_006230251.1 | 4 - 5 | 914 | 96 | ||
GenBank mRNA | BC101866.1 | - | 4 - 5 | 641 | 7938969 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |