Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019158.2 | NP_062031.1 | 3 - 4 | 517 | 81 | |
XM_006230187.2 | XP_006230249.1 | 3 - 4 | 564 | 81 | ||
GenBank mRNA | AB005547.1 | - | 3 - 4 | 516 | 81 | |
AF007775.1 | - | 3 - 4 | 496 | 81 | ||
BC081812.1 | - | 3 - 4 | 517 | 7114305 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |