Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asphd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759931.1 | XP_008758153.1 | 1 - 2 | 1647 | 78 | |
XM_215083.7 | XP_215083.2 | 1 - 2 | 1647 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |