Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106297.2 | NP_001099767.1 | 2 - 3 | 252 | 94 | |
XM_006230251.2 | XP_006230313.1 | 3 - 4 | 2014 | 94 | ||
XM_006230252.2 | XP_006230314.1 | 3 - 4 | 2022 | 94 | ||
XM_008759879.1 | XP_008758101.1 | 3 - 4 | 2028 | 94 | ||
GenBank mRNA | BC168174.1 | - | 2 - 3 | 252 | 9032868 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |