Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004222.1 | NP_001004222.1 | 3 - 4 | 333 | 90 | |
XM_006229933.2 | XP_006229995.1 | 3 - 4 | 351 | 90 | ||
GenBank mRNA | BC079174.1 | - | 3 - 4 | 333 | 7130075 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |