Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106298.1 | NP_001099768.1 | 2 - 3 | 430 | 100 | |
XM_006230256.2 | XP_006230318.1 | 2 - 3 | 323 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |