Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106299.1 | NP_001099769.1 | 2 - 3 | 200 | 102 | |
GenBank mRNA | BC157822.1 | - | 2 - 3 | 124 | 8374205 | 102 |
FQ225822.1 | - | 2 - 3 | 121 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |