Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191877.1 | NP_001178806.1 | 5 - 6 | 664 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |