Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108990.1 | NP_001102460.1 | 4 - 5 | 377 | 135 | |
XM_006241337.2 | XP_006241399.1 | 14 - 15 | 1725 | 135 | ||
XM_008765359.1 | XP_008763581.1 | 13 - 14 | 1257 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |