Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017458.1 | NP_001017458.1 | 6 - 7 | 1024 | 145 | |
XM_006241174.2 | XP_006241236.1 | 7 - 8 | 1086 | 145 | ||
GenBank mRNA | BC092644.1 | - | 6 - 7 | 1024 | 7318345 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |