Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134887.1 | NP_001128359.1 | 5 - 6 | 716 | 91 | |
XM_006241203.2 | XP_006241265.1 | 5 - 6 | 717 | 91 | ||
XM_006241204.2 | XP_006241266.1 | 5 - 6 | 718 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |