Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241426.2 | XP_006241488.1 | 7 - 8 | 2485 | 87 | |
XM_006241428.2 | XP_006241490.1 | 6 - 7 | 2238 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |