Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108081.1 | NP_001101551.1 | 6 - 7 | 633 | 90 | |
XM_006241243.1 | XP_006241305.1 | 6 - 7 | 642 | 90 | ||
GenBank mRNA | BC166860.1 | - | 6 - 7 | 609 | 9101114 | 90 |
FQ220136.1 | - | 3 - 4 | 278 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |