Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012886.2 | NP_037018.2 | 2 - 3 | 495 | 89 | |
XM_006241162.2 | XP_006241224.1 | 2 - 3 | 524 | 89 | ||
GenBank mRNA | BC097335.1 | - | 2 - 3 | 495 | 7443686 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |