Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055430.5 | XP_001055430.4 | 4 - 5 | 914 | 106 | |
XM_006241414.2 | XP_006241476.1 | 5 - 6 | 508 | 106 | ||
XM_008765427.1 | XP_008763649.1 | 4 - 5 | 934 | 106 | ||
XM_008765428.1 | XP_008763650.1 | 5 - 6 | 558 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |