Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191781.1 | NP_001178710.1 | 4 - 5 | 601 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |