Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759836.1 | XP_008758058.1 | 56 - 57 | 11522 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |