Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024885.1 | NP_001020056.1 | 8 - 9 | 1011 | 82 | |
GenBank mRNA | BC097480.1 | - | 8 - 9 | 1011 | 7458611 | 82 |
FQ215669.1 | - | 8 - 9 | 713 | 82 | ||
FQ216256.1 | - | 8 - 9 | 715 | 82 | ||
FQ218235.1 | - | 8 - 9 | 726 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |