Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108920.1 | NP_001102390.1 | 4 - 5 | 786 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |