Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039208.2 | NP_001034297.2 | 10 - 11 | 1660 | 100 | |
GenBank mRNA | BC110048.1 | - | 7 - 8 | 2493 | 7315879 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |