Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039208.2 | NP_001034297.2 | 4 - 5 | 943 | 87 | |
GenBank mRNA | BC110048.1 | - | 4 - 5 | 928 | 7315879 | 87 |
BU671623.1 | - | 1 - 2 | 126 | 87 | ||
CA339235.1 | - | 4 - 5 | 516 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |